missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ EZ-Link™ Maleimide-PEG2-Biotin, No-Weigh™ Format
Biotinylate reduced cysteines with this short, maleimide-activated, sulfhydryl-reactive biotinylation reagent containing a 2-unit PEG spacer arm to enhance water solubility.
£269.00 - £379.00
Specifications
Product Type | Maleimide-PEG2-Biotin |
---|---|
Product Line | EZ-Link™ |
Cell Permeability | Cell-Impermeant |
Chemical Reactivity | Thiol |
Reactive Moiety | Maleimide |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10519873
|
Thermo Scientific™
21901BID |
50 mg |
£379.00
50mg |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15980186
|
Thermo Scientific™
A39261 |
10 x 2 mg |
£269.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
Description
Thermo Scientific™ EZ-Link Maleimide-PEG2-Biotin is a mid-length, maleimide-activated, sulfhydryl-reactive biotinylation reagent that contains a 2-unit ethylene glycol in its spacer arm for increased water-solubility characteristics.
Highlights:
- Water-soluble; contains polyethylene glycol (PEG) spacer arm
- Reacts with sulfhydryl (-SH) groups at acidic to neutral pH
- Transfers hydrophilic PEG spacer arm to final conjugate
- Forms a thioether bond
Specifications
Maleimide-PEG2-Biotin | |
Cell-Impermeant | |
Maleimide | |
Biotin |
EZ-Link™ | |
Thiol | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water |
For Research Use Only. Not for use in diagnostic procedures.