Learn More
Thermo Scientific™ EZ-Link™ Sulfo-NHS-Biotin
Thermo Scientific™ EZ-Link™ Sulfo-NHS-Biotin is a short-chain, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.
Brand: Thermo Scientific™ A39256
149.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Additional Details : CAS Number : 119616-38-5 Weight : 0.05200kg
Description
Label antibodies, proteins and other molecules that have primary amines with Thermo Scientific™ EZ-Link Sulfo-NHS-Biotin, a short-chain, water-soluble biotinylation reagent.
- Protein labeling - biotinylate antibodies to facilitate immobilization, purification or detection using streptavidin resins or probes
- Cell surface labeling - biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
- Amine-reactive - reacts with primary amines (-NH2), such as lysine side-chains or the amino-termini of polypeptides
- Soluble - charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
- Irreversible - forms permanent amide bonds; spacer arm cannot be cleaved
- Very short - spacer arm (total length added to target) is 13.5 angstroms; it consists of the native biotin valeric acid group only.
Specifications
10 x 1 mg | |
10 x 1 mg | |
Chemical Labeling | |
EZ-Link™ | |
Amine | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Short |
Sulfo-NHS-Biotin | |
Store desiccated at -20°C. | |
Qualified for Ambient or Wet Ice | |
Cell-Impermeant | |
Sulfo-NHS Ester | |
Biotin |
For Research Use Only. Not for use in diagnostic procedures.