missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ EZ-Link™ Sulfo-NHS-Biotin
Thermo Scientific™ EZ-Link™ Sulfo-NHS-Biotin is a short-chain, water-soluble biotinylation reagent for labeling antibodies, proteins and other molecules that have primary amines.
£168.00 - £269.00
Specifications
Product Type | Sulfo-NHS-Biotin |
---|---|
Product Line | EZ-Link™ |
Cell Permeability | Cell-Impermeant |
Chemical Reactivity | Amine |
Solubility | DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17872556
|
Thermo Scientific™
A39256 |
10 x 1 mg |
£168.00
Pack of 10 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15940737
|
Thermo Scientific™
A39256 |
10 x 1 mg |
£168.00
Pack of 10 |
In Stock
Log in to see stock. |
|||||
11851185
|
Thermo Scientific™
21217 |
50 mg |
£269.00
50mg |
In Stock
Log in to see stock. |
|||||
Description
Label antibodies, proteins and other molecules that have primary amines with Thermo Scientific™ EZ-Link Sulfo-NHS-Biotin, a short-chain, water-soluble biotinylation reagent.
- Protein labeling - biotinylate antibodies to facilitate immobilization, purification or detection using streptavidin resins or probes
- Cell surface labeling - biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
- Amine-reactive - reacts with primary amines (-NH2), such as lysine side-chains or the amino-termini of polypeptides
- Soluble - charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
- Irreversible - forms permanent amide bonds; spacer arm cannot be cleaved
- Very short - spacer arm (total length added to target) is 13.5 angstroms; it consists of the native biotin valeric acid group only.
Specifications
Sulfo-NHS-Biotin | |
Cell-Impermeant | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Short |
EZ-Link™ | |
Amine | |
Biotin |
For Research Use Only. Not for use in diagnostic procedures.