Learn More
Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin, No Weigh Format
Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin No-Weigh™ Format is a water-soluble, NHS-ester biotinylation reagent whose spacer arm includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines.
Brand: Thermo Scientific™ 21331
397.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
Thermo Scientific™ EZ-Link Sulfo-NHS-SS-Biotin enables simple and efficient biotin labeling of antibodies, proteins and any other primary amine-containing biomolecules in solution.
Highlights:
- Cleavable analog of Sulfo-NHS-LC-Biotin
- Can remove biotin label by cleaving spacer disulfide bond with reducing agents, such as DTT
- Ideal for affinity purification experiments that require efficient elution from immobilized avidin supports (cleaving disulfide bond releases proteins)
- Long spacer arm reduces steric hindrance for binding interactions
Specifications
EZ-Link Sulfo-NHS-SS-Biotin | |
100 mg | |
EZ-Link™ | |
Amine | |
DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water | |
Mid-length, Cleavable |
Sulfo-NHS-SS-Biotin | |
Store at 4°C. | |
Cell-Impermeant | |
Active Ester, Succinimidyl Ester, Sulfo-NHS Ester | |
Biotin | |
Biotin & Analogs |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.