Learn More
Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin, No Weigh Format
Thermo Scientific™ EZ-Link™ Sulfo-NHS-SS-Biotin No-Weigh™ Format is a water-soluble, NHS-ester biotinylation reagent whose spacer arm includes a cleavable disulfide bond for reversible labeling of proteins and cell surface primary amines.
£150.00 - £483.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15925521
|
Thermo Scientific™
A39258 |
10 x 1 mg |
£214.00
10mg |
In Stock
Log in to see stock. |
|||||
11811205
|
Thermo Scientific™
21331 |
100 mg |
£483.00
100mg |
In Stock
Log in to see stock. |
|||||
10361554
|
Thermo Scientific™
21328 |
8 x 1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Thermo Scientific™ EZ-Link Sulfo-NHS-SS-Biotin enables simple and efficient biotin labeling of antibodies, proteins and any other primary amine-containing biomolecules in solution.
- Protein labeling - biotinylate antibodies to facilitate immobilization, purification or detection
- Cell surface labeling - biotinylates only surface proteins of whole cells because the negatively charged reagent does not permeate cell membranes
- Amine-reactive - reacts with primary amines (-NH2), such as lysine side-chains, or the amino-termini of polypeptides
- Cleavable - disulfide bond in spacer arm allows the biotin label to be removed using reducing agents such as DTT; only a small sulfhydryl group remains attached to the molecule
- Soluble - charged sulfo-NHS group increases reagent water solubility compared to ordinary NHS-ester compounds
- Medium length - spacer arm (total length added to target) is 24.3 angstroms; it consists of the native biotin valeric acid group extended by a 7-atom chain
Specifications
Amine |
For Research Use Only. Not for use in diagnostic procedures.