missing translation for 'onlineSavingsMsg'
Learn More

Julabo™ F Series Recirculating Cooling Baths with Air Compressor

Require very little laboratory space and have very low procurement costs. . Julabo™ F Series Recirculating Cooling Baths with Air Compressor are a great way to replace costly systems based on running tap water and are ideal for basic cooling tasks.

Brand:  Julabo™ 9620100

4213.38 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 15214373

  • £5340.00 / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Environmentally friendly operation with low energy consumption
  • Compact design
  • Splashproof membrane keypad with LED temperature display
  • Straightforward filling and draining
  • Filling level indicator
  • May be used with water, water/glycol, Julabo Thermal G
Specifications

Specifications

9.5 L
0.35kW at 0°C, 0.55kW at 5°C, 0.7kW at 10°C, 1kW at 20°C (Medium Ethanol)
For simple cooling tasks
14.5 psi
R134A
±0.5°C
37.59 cm
60.78 kg
115/230 V 60 Hz
60 Hz
CE
23 L/min.
0°C to 40°C
F1000
±0.5°C
49.02 cm
64 cm
45 kg
115/230 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.