missing translation for 'onlineSavingsMsg'
Learn More

Falcon™ Sterile Sample Containers

Made of clear polypropylene for strength and visibility. Falcon™ Sterile Sample Containers are ideal for specimen collection.

Brand:  Falcon™ 354020

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10153862

  • £305.93 / Pack of 500
Estimated Shipment: 11-12-2024
to see stock.

Description

Description

  • Sterile, disposable polypropylene containers provide secure sample containment
  • Available in 4 1/2 oz. (110 mL) and 8 oz. (220 mL) sizes both with and without matching lids
  • Feature molded-in graduations for easy measurements
  • Graduated in ounces and milliliters to provide convenient, single-use, timesaving containers for collection, transportation, and storage of a wide variety of specimens
  • Inert and chemically resistant to commonly used laboratory reagents at room temperature
  • Features

    Sterile
  • 4 1/4 oz. graduations from 1/2 oz. to 4 1/2 oz. in 1/4 oz. increments and from 20 mL to 110 mL in 10 mL increments
  • 8 oz. size graduated from 1/4 oz. to 8 oz. in 1/4 oz. increments and from 20 mL to 220 mL in 10 mL increments
Specifications

Specifications

No
Polypropylene
220 mL
20/Pk.
Yes
Sterile
500/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers