missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Falcon™ Sterile Sample Containers
Made of clear polypropylene for strength and visibility. Falcon™ Sterile Sample Containers are ideal for specimen collection.
£87.68 - £337.00
Product Code | Brand | Capacity (Metric) | Closure Included | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Included | Price | Quantity & Availability | ||||
10100881
|
Falcon™
354015 |
220 mL | Yes |
£91.91
Pack of 100 |
Estimated Shipment: 15-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10153862
|
Falcon™
354020 |
220 mL | No |
£305.93
Pack of 500 |
Estimated Shipment: 11-12-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10002922
|
Falcon™
354014 |
110 mL | No |
£337.00
Pack of 500 |
Estimated Shipment: 11-12-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10190871
|
Falcon™
354013 |
110 mL | Yes |
£87.68
Pack of 100 |
Estimated Shipment: 06-12-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Sterile, disposable polypropylene containers provide secure sample containment
- Available in 4 1/2 oz. (110 mL) and 8 oz. (220 mL) sizes both with and without matching lids
- Feature molded-in graduations for easy measurements
- Graduated in ounces and milliliters to provide convenient, single-use, timesaving containers for collection, transportation, and storage of a wide variety of specimens
- Inert and chemically resistant to commonly used laboratory reagents at room temperature
Features
Sterile- 4 1/4 oz. graduations from 1/2 oz. to 4 1/2 oz. in 1/4 oz. increments and from 20 mL to 110 mL in 10 mL increments
- 8 oz. size graduated from 1/4 oz. to 8 oz. in 1/4 oz. increments and from 20 mL to 220 mL in 10 mL increments
Specifications
Yes | |
Polypropylene | |
Sterile |