missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Falcon™ Sterile Sample Containers
Made of clear polypropylene for strength and visibility. Falcon™ Sterile Sample Containers are ideal for specimen collection.
Brand: Falcon™ 354013
This item is not returnable.
View return policy
Description
- Sterile, disposable polypropylene containers provide secure sample containment
- Available in 4 1/2 oz. (110 mL) and 8 oz. (220 mL) sizes both with and without matching lids
- Feature molded-in graduations for easy measurements
- Graduated in ounces and milliliters to provide convenient, single-use, timesaving containers for collection, transportation, and storage of a wide variety of specimens
- Inert and chemically resistant to commonly used laboratory reagents at room temperature
Features
Sterile- 4 1/4 oz. graduations from 1/2 oz. to 4 1/2 oz. in 1/4 oz. increments and from 20 mL to 110 mL in 10 mL increments
- 8 oz. size graduated from 1/4 oz. to 8 oz. in 1/4 oz. increments and from 20 mL to 220 mL in 10 mL increments
Specifications
Yes | |
Polypropylene | |
110 mL | |
1/Pk. |
Yes | |
Sterile | |
100/Cs. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction