missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM71F2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 24286-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Especificaciones
FAM71F2 | |
Polyclonal | |
Unconjugated | |
FAM71F2 | |
FAM137B, FAM71F2, Protein FAM137B, Protein FAM71F2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
245884, 346653 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.66 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6NXP2 | |
FAM71F2 | |
FAM71F2 Fusion Protein Ag16874 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |