missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FAM71F2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | FAM71F2 |
---|---|
Concentration | 0.66 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16802765
|
Proteintech
24286-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16892755
|
Proteintech
24286-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
FAM71F2 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q6NXP2 | |
245884, 346653 | |
FAM71F2 Fusion Protein Ag16874 | |
Primary | |
-20°C | |
FAM71F2 |
0.66 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
FAM137B, FAM71F2, Protein FAM137B, Protein FAM71F2 | |
FAM71F2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |