missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FastDigest SmaI

The FastDigest SmaI restriction enzyme recognizes CCC^GGG sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (isoschizomers: TspMI, XmaCI; neoschizomer: XmaI).

Brand:  Thermo Scientific™ FD0663

17.71 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 10110890

  • £26.57 / 100 reactions

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

FastDigest SmaI (100 μL), 10X FastDigest Buffer (1.0 mL), 10X FastDigest Green Buffer (1.0 mL), Store at –20°C.
Sma I
Yes
No
Restriction Enzyme
FastDigest™
65°C
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
10X FastDigest Buffer/FastDigest Green Buffer
37°C
100 μL (100 Reactions)
Traditional Cloning
Blunt
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2821329 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberFD0663
Lot Number2903859 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberFD0663
Lot Number2983417 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberFD0663
Lot Number2943824 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberFD0663
Lot Number2966170 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberFD0663
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ FastDigest SmaI > Quantity: 100 μL (100 Reactions)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.