missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals FastGlycoProtein™ Isolation Kit, WGA Resin

Used to isolate glycoprotein from complex protein mixtures using the lectin wheat germ agglutinin (WGA) immobilized on agarose.

Brand:  MP Biomedicals 116550900

Additional Details : Weight : 0.12000kg

Product Code. 15712462

  • £988.98 / 1 set
Estimated Shipment: 05-09-2024
to see stock.
Add to basket

Description

Description

  • Isolates glycoprotein from complex protein mixtures including animal and plant tissue, cultured cells, including microbe and insect cells, using the lectin wheat germ agglutinin (WGA) immobilized on agarose.
  • Works with: QuickPrep (24x2 mL); HiPrep (48x2 mL); CoolPrep (24x2 mL); QuickFlex (96x2 mL); ConeFlex Legacy (allows all FP24 adapters on FP96).
  • Alternate name: FastGlycoProtein™ Isolation Kit WGA Resin.
  • Capacity: Samples containing up to 1.5 mg protein .
  • Components: Lysing Matrix A 10 x 2 mL Tubes, Garnet matrix and 1/4 inch ceramic spheres, WGA Lectin Resin 1.1 mL settled, as slurry, Binding/Wash Buffer. 5X solution 6.5 mL, Elution Solution 5 mL, Spin Filter with caps 10 each, Catch Tubes 20 each.
  • Format: Bead beating or chemical lysis.
  • Hardness: Zirconium sphere, Rockwell A 80-84, Vickers 1050; Garnet Flakes, Mohs 7.5-8.
  • Kit/Assay type: Affinity resin with spin columns.
  • Required materials not supplied: FastPrep™ Instrument or alternative high speed disruptor; Microcentrifuge capable of 10,000 xg rcf that can freely spin 2.0 mL tubes; Microcentrifuge tubes (2.0 mL and 1.5 mL); Rotator or low-speed vortex; sterile water.
  • Molecule enriched: Glycoprotein.
  • Research areas: Immune regulation, inflammation, cell-to-cell adhesion and contact inhibition, cell signaling, protection against proteolytic degradation, and other biological processes
  • Sample throughput: 1-10 x 2 mL tubes.
  • Research Use Only (RUO).
Specifications

Specifications

Glycoprotein Isolation Kit
1 Kit, throughput of 1-10 2 mL Tubes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers