Learn More
Thermo Scientific™ FastDigest NotI
The FastDigest NotI restriction enzyme recognizes GC^GGCCGC sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: CciNI).
Brand: Thermo Scientific™ FD0596
200.88 GBP valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
FastDigest enzymes are an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.
The universal buffer allows rapid single-, double- or multiple DNA digestion within 5-15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. DNA modifying enzymes, such as Klenow Fragment, T4 DNA Ligase, alkaline phosphatases and T4 DNA Polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.
For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.
Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.
Thermo Scientific FastDigest NotI restriction enzyme recognizes GC^GGCCGC site and cuts best at 37°C in 5–15 minutes using universal FastDigest Buffer. Isoschizomers: CciNI.
Thermo Scientific FastDigest NotI is one of an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.
The universal buffer allows rapid single-, double-, or multiple DNA digestion within 5–15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. See Reaction Conditions for FastDigest Enzymes for a table of enzyme activity, heat inactivation and incubation times for this and other FastDigest restriction enzymes. DNA modifying enzymes, such as Klenow Fragment (Cat. No. EP0051), T4 DNA Ligase (Cat. No. EL0014), alkaline phosphatases and T4 DNA Polymerase (Cat. No. EP0061) all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.
For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.
Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.
Features
• 100% activity of all FastDigest enzymes in the universal buffer
• 100% buffer compatibility with downstream applications
• Complete digestion in 5–15 minutes
• Direct loading on gels
• No star activity
• 176 FastDigest enzymes available
Applications
• Molecular cloning
• Restriction site mapping
• Genotyping
• Southern Blot
• Restriction fragment length polymorphism (RFLP)
• SNP analysis
Note: The FastDigest Green Buffer offers the same high performance in DNA digestion and downstream applications as the colorless FastDigest Buffer. For applications that require product analysis by fluorescence excitation (e.g. concentration measurements in UV light) the colorless FastDigest Buffer is recommended.
For methylation sensitivity, refer to product specifications.

Specifications
• FastDigest NotI (250 μL) • 10X FastDigest Buffer (1.0 mL) • 10X FastDigest Green Buffer (1.0 mL) Store at –20°C. |
|
Not I | |
Yes | |
No | |
Restriction Enzyme | |
FastDigest™ | |
80°C |
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive | |
10X FastDigest Buffer/FastDigest Green Buffer | |
37°C | |
250 μL (250 Reactions) | |
Traditional Cloning | |
5' protruding ends |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.