missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneRuler 100 bp DNA Ladder, ready-to-use
GREENER_CHOICE

Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.

£72.25 - £307.00

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Product Type DNA Ladder
Ready to Load Yes
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10328179
GREENER_CHOICE
View Documents
Thermo Scientific™
SM0243
50 μg, 100 applications
£72.25
50µg
Please to purchase this item. Need a web account? Register with us today!
11883953
GREENER_CHOICE
View Documents
Thermo Scientific™
SM0244
5 x 50 μg, 500 applications
£307.00
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Description

Description

GeneRuler 100 bp DNA Ladder, ready-to-use 100 to 1000 bp is recommended for sizing and approximate quantification of a wide range of double-stranded DNA fragments on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments.

GeneRuler 100 bp DNA Ladder, ready-to-use, is premixed with 6X DNA Loading Dye for direct loading on gel.

The ladder is supplied with 6X DNA Loading Dye.

Highlights

  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Can be directly loaded and are stable at room temperature for 6 months
  • Supplied with loading dye for sample DNA
Specifications

Specifications

6X
Xylene Cyanol FF, Bromophenol Blue, Orange G
Yes
Agarose Gels
GeneRuler
Agarose
DNA Ladder
100 to 1000 bp
Sustainable packaging
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 6 Months

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ GeneRuler 100 bp DNA Ladder, ready-to-use

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.