Learn More
Thermo Scientific™ GeneRuler 1 kb DNA Ladder, ready-to-use
GREENER_CHOICE
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels.
Brand:  Thermo Scientific™ SM0314
34.00 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
 View more versions of this product
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10809360
  • £50.75 / 50µg
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
GeneRuler 1 kb DNA Ladder, ready-to-use, is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels. The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler 1 kb DNA Ladder, ready-to-use, is premixed with 6X DNA Loading Dye for direct loading on gel.
The ladder is supplied with 6X DNA Loading Dye.
Highlights
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Can be directly loaded and are stable at room temperature for 6 months
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6X
Xylene Cyanol FF, Bromophenol Blue, Orange G
Yes
Agarose Gels
GeneRuler
Agarose
DNA Ladder
250 to 10000 bp
Sustainable packaging
50 μg, 100 applications