Learn More
Thermo Scientific™ KpnI, HC (50 U/μL)
The KpnI restriction enzyme recognizes GGTAC^C sites and cuts best at 37°C in its own unique buffer (Isoschizomers: Asp718I).
Brand: Thermo Scientific™ ER0522
215.28 GBP valid until 2025-03-28
Use promo code "24107" to get your promotional price.
Description
Lambda DNA digested with KpnI, 0.7% agarose, 2 cleavage sites
conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.
5'..G G T A C▵C...3'
3'..C▵C A T G G...5'
Conditions for 100% Activity:
- 1X Buffer KpnI: 10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 0.02% Triton X-100 and 0.1mg/mL BSA
- Incubate at 37°C
Storage Buffer:
- KpnI is supplied in: 10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol
Methylation Effects:
- Dam: never overlaps —no effect
- Dcm: may overlap —no effect
- CpG: may overlap —no effect
- EcoKI: never overlaps —no effect
- EcoBI: never overlaps —no effect
Digestion of Agarose-embedded DNA:
- Minimum 5U of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours
Note:
High glycerol (>5%) concentrations, pH>8.0 or a large excess of enzyme may result in star activity.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
10 U/μL | |
Kpn I | |
Yes | |
No | |
Restriction Enzyme |
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive | |
Unique Buffer (10X Buffer KpnI) | |
37°C | |
5 x 4,000 units | |
Traditional Cloning |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
New Offer Shop Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.