missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RNase I (10 U/μL)

Ribonuclease I (RNase I) is an endoribonuclease that hydrolyzes single-stranded RNA to nucleoside 3'-monophosphates.

Brand:  Thermo Scientific™ EN0602

206.50 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10374290

This item is not returnable. View return policy

Description

Description

Ribonuclease I (RNase I), an endoribonuclease, preferentially hydrolyzes single-stranded RNA to nucleoside 3'-monophosphates via nucleoside 2', 3'-cyclic monophosphate intermediates.

The enzyme does not require any metal ions for activity.

Highlights

  • Stable and active under a wide variety of reaction conditions
  • Can be heat inactivated in 30 minutes at 100°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 U/μL
RNase
5000 U
RNase I
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.