Learn More
Thermo Scientific™ T4 DNA Polymerase (5 U/μL)
Ensure accuracy in DNA synthesis with template-dependent T4 DNA Polymerase that catalyzes the 5'-3' synthesis from primed single-stranded DNA.
£45.77 - £188.00
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
10680281
|
Thermo Scientific™
EP0061 |
1mL of 5X Reaction Buffer |
£45.77
100 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10531051
|
Thermo Scientific™
EP0062 |
2 x 1mL of 5X Reaction Buffer |
£188.00
500 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
T4 DNA Polymerase, a template-dependent DNA polymerase, catalyzes 5'-3' synthesis from primed single-stranded DNA. The enzyme has a 3'-5' exonuclease activity, but lacks 5'-3' exonuclease activity.
Highlights
- Stronger 3'-5' exonuclease activity on single-stranded than on double-stranded DNA and greater (more than 200 times) than DNA polymerase I, E. coli,and Klenow fragment
- Active in Thermo Scientific restriction enzyme, PCR, RT and T4 DNA Ligase buffers
Specifications
T4 DNA Polymerase |