missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Uracil-DNA Glycosylase (1 U/μL)
Uracil-DNA Glycosylase (UDG, UNG) catalyzes the hydrolysis of the N-glycosylic bond between uracil and sugar in DNA and prevents carry-over of DNA in PCR reactions.
£42.64 - £176.00
Specifications
Concentration | 1 U/μL |
---|---|
Content And Storage | Uracil-DNA glycolase (1 U/μL) and 10X Reaction Buffer. Store at -20°C. |
Enzyme | Uracil-DNA Glycosylase |
Compatible Buffer | 10X Reaction Buffer |
Product Type | Uracil DNA Glycosylase |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10621041
|
Thermo Scientific™
EN0361 |
200 U |
£42.64
200 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10631041
|
Thermo Scientific™
EN0362 |
5 x 200 U |
£176.00
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Uracil-DNA Glycosylase (UDG, UNG) catalyzes the hydrolysis of the N-glycosylic bond between uracil and sugar, leaving an apyrimidinic site in uracil-containing single or double-stranded DNA. The enzyme shows no activity on RNA.
Highlights
- Active in Thermo Scientific buffers for restriction enzymes and thermophilic polymerases
Specifications
1 U/μL | |
Uracil-DNA Glycosylase | |
Uracil DNA Glycosylase |
Uracil-DNA glycolase (1 U/μL) and 10X Reaction Buffer. Store at -20°C. | |
10X Reaction Buffer |
For Research Use Only. Not for use in diagnostic procedures.