missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, qualified, Brazil

Virus and Mycoplasma Tested. Supplied with a Certificate of Analysis detailing Q.C results.

Brand:  Gibco™ 10270106

Code : 50

Additional Details : Weight : 0.52560kg

 View more versions of this product

Product Code. 11573397

  • £367.00 / 500mL
Estimated Shipment: 29-07-2024
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Sera Category: Standard
Origin: E.U. Approved (South American)
Virus and Mycoplasma Tested. Supplied with a Certificate of Analysis detailing Q.C results.
Specifications

Specifications

Basic cell culture, specialty research, and specific assays
Store in freezer (–5 to –30°C)
E.U. Approved (South American)
Bottle
500 mL
Cattle/Bovine
Liquid
EU-Approved
Fetal Bovine Serum, E.U. Approved (South American Origin), 500mL, Liquid, –5 to –30°C Storage Temperature, Fetal, Cattle/Bovine Species, Standard (Sterile-Filtered) Serum Treatment, Dry Ice, Virus and Mycoplasma Tested
Liquid
Fetal Bovine Serum
Sterile-filtered
Virus, Mycoplasma
Basic cell culture, specialty research, and specific assays
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers