Learn More
Gibco™ Fetal Bovine Serum, qualified, heat inactivated, United States

Important Update: We've recently introduced Premium Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Qualified US origin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
£109.00 - £592.00
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | Fetal |
Country of Origin | United States |
Form | Liquid |
Product Type | Fetal Bovine Serum |
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Format | Price | Quantity & Availability | ||||||
15828947
![]() |
Gibco™
A3840102 |
10 x 50 mL | One Shot | N/A | N/A | N/A | |||||
11570516
![]() |
Gibco™
16140063 |
100 mL | Bottle | N/A | N/A | N/A | |||||
11580516
![]() |
Gibco™
16140071 |
500 mL | Bottle | N/A | N/A | N/A | |||||
Description
Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Value Plus (Performance)
- Origin: United States.
- Endotoxin level: ≤10 EU/mL
- Hemoglobin level: ≤25 mg/dL
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
United States | |
Fetal Bovine Serum | |
≤10 EU/mL | |
Qualified |
Fetal | |
Liquid | |
Heat-inactivated | |
≤25 mg/dL | |
Frozen |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.