Learn More
Fexofenadine-d6, MedChemExpress
MedChemExpress Fexofenadine D6 (MDL-16455 D6) is deuterium labeled is Fexofenadine, which is an antihistamine pharmaceutical agent.
Brand: Medchem Express HY-B0801S
This item is not returnable.
View return policy
Description
Fexofenadine D6 (MDL-16455 D6) is deuterium labeled is Fexofenadine, which is an antihistamine pharmaceutical agent.Specifications
Fexofenadine-d6 | |
1 mg | |
Research | |
MDL-16455-d6 Terfenadine carboxylate-d6 | |
507.69 | |
98.0% | |
Neuroscience-Neuromodulation |
548783-71-7 | |
White | |
C32H33D6NO4 | |
OC(C1=CC=CC=C1)(C2=CC=CC=C2)C3CCN(CCCC(C4=CC=C(C(C(O)=O)(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C4)O)CC3 | |
507.69 | |
Research | |
Powder |