missing translation for 'onlineSavingsMsg'
Learn More

Fexofenadine-d6, MedChemExpress

MedChemExpress Fexofenadine D6 (MDL-16455 D6) is deuterium labeled is Fexofenadine, which is an antihistamine pharmaceutical agent.

POA - £744.15

Specifications

Chemical Name or Material Fexofenadine-d6
CAS 548783-71-7
Color White
Purity Grade Notes Research
Molecular Formula C32H33D6NO4
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16432279
Encompass_Preferred
View Documents
Medchem Express
HY-B0801S
1 mg
£228.69
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16418628
Encompass_Preferred
View Documents
Medchem Express
HY-B0801S
10 mg
POA
10mg
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

16401078
Encompass_Preferred
View Documents
Medchem Express
HY-B0801S
5 mg
£744.15
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fexofenadine D6 (MDL-16455 D6) is deuterium labeled is Fexofenadine, which is an antihistamine pharmaceutical agent.
Specifications

Specifications

Fexofenadine-d6
White
C32H33D6NO4
OC(C1=CC=CC=C1)(C2=CC=CC=C2)C3CCN(CCCC(C4=CC=C(C(C(O)=O)(C([2H])([2H])[2H])C([2H])([2H])[2H])C=C4)O)CC3
507.69
Research
Powder
548783-71-7
Research
MDL-16455-d6 Terfenadine carboxylate-d6
507.69
98.0%
Neuroscience-Neuromodulation
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.