Learn More
Invitrogen™ FGF19 Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ EHFGF19X5
Description
The Human Fibroblast Growth Factor 19 (Hu FGF19) ELISA quantitates Hu FGF19 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu FGF19. Principle of the method The Human FGF19 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Fibroblast growth factor-1 (FGF-1), also designated acidic FGF, and fibroblast growth factor-2 (FGF-2), also designated basic FGF, are members of a family of growth factors that stimulate proliferation of cells of mesenchymal, epithelial and neuroectodermal origin. Additional members of the FGF family include the oncogenes FGF-3 (Int2) and FGF-4 (hst/Kaposi), FGF-5, FGF-6, FGF-7 (KGF), FGF-8 (AIGF), FGF-9 (GAF) and FGF-10-FGF-23. Members of the FGF family share 30-55% amino acid sequence identity and similar gene structure, and are capable of transforming cultured cells when overexpressed in transfected cells. Cellular receptors for FGFs are members of a second multigene family including four tyrosine kinases, designated Flg (FGFR-1), Bek (FGFR-L), TKF and FGFR-3.Specifications
O95750 | |
30 pg/mL | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
5 x 96 assays | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
9965 | |
<12% | |
Assay Diluent Concentrate, Adhesive Plate Covers, Standard, Pre-coated 96 well plates, SAV-HRP, Biotinylated Detection Antibody, Stop Solution, Wash Buffer, Chromogen | |
5 x 96 Tests | |
Neurobiology | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
32.77 pg/mL to 8000 pg/mL | |
30 pg/mL to 8000 pg/mL | |
Biotin | |
Growth Factors & Receptors | |
This ELISA antibody pair detects human FGF-19. Other species not determined yet. | |
Human | |
Colorimetric Microplate Reader | |
FGF19 | |
<10% | |
HRP | |
RUO | |
Plasma, 50 μ;L; Serum, 50 μ;L; Supernatant, 100 μ;L | |
UNQ334/PRO533 | |
1 hr. 20 min. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.