Learn More
Fisherbrand™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Organize vial storage within cryo boxes with these fiberboard box dividers.
Brand:  Fisherbrand™ 5958
28.35 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
Product Code. 11986924
  • £29.84 / Pack of 12
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • Use with fiberboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vials (100)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers >