Learn More
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Organize vial storage within cryo boxes with these fiberboard box dividers.
Brand:  Thermo Scientific™ AVL5959
 View more versions of this product
Product Code. 12744388
  • £11.70 / Each
Estimated Shipment: 18-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
  • Use with fiberboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
TRUSTED_SUSTAINABILITY
Specifications
Specifications
12 Pack
49 (7 x 7 Grid)
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers > Holds 49 x 16mm vials; Grid size: 7 x 7; Pack of 12