Learn More
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Organize vial storage within cryo boxes with these fiberboard box dividers.
£3.90 - £29.56
Products 6
Product Code Brand Holds Quantity Price Quantity & Availability  
Product Code Brand Holds Quantity Price Quantity & Availability  
12744388
View Documents
Thermo Scientific™
AVL5959
49 (7 x 7 Grid) 12 Pack
£11.70
Each
Estimated Shipment: 14-08-2025
to see stock.
Add to basket
15810530
View Documents
Thermo Scientific™
5960
64 (8 x 8 Grid) 12 Pack
£28.83
Pack of 12
Estimated Shipment: 21-07-2025
to see stock.
Add to basket
11849490
View Documents Promotion Details
Thermo Scientific™
AVL6212
81 (9 x 9 Grid) 12 Pack
£29.56
Pack of 12
In Stock
to see stock.
Edge
Add to basket
15907976
View Documents
Thermo Scientific™
820025
25 (5 x 5 Grid) Each
£3.97
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
15821152
View Documents Promotion Details
Thermo Scientific™
820049
49 (7 x 7 Grid) Each
£3.97
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
11329341
View Documents Promotion Details
Thermo Scientific™
820081
81 (9 x 9 Grid) Each
£3.90
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
  • Use with fiberboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers