Learn More
Thermo Scientific™ Fiberlite™ F21-48 x 2 Fixed-Angle Rotor
Support micro-volume protocols using the fixed angle rotor, provided with a biocontainment lid.
Brand: Thermo Scientific™ 75003664
2297.10 GBP valid until 2025-05-09
Use promo code "24143" to get your promotional price.
Description
Support micro-volume protocols using the Thermo Scientific™ Fiberlite™ F21-48 x 2 Fixed-Angle Rotor.
Features:
Fiberlite Rotor:
- Reduces run-times with higher G-forces achieved with lightweight carbon fiber rotors
- Ensures structural integrity with corrosion- and fatigue-resistant design
- Backed by a 15-year warranty
Ordering Information:
Tube adapters are sold separately in sets of 24; O-ring Refresher Kit (021-489-021) is also available. Contact your sales representative for information.
Compatible with:
Thermo Scientific Sorvall™ Legend™ X1, X3, Sorvall ST 40, Multifuge™ X1, X3, Megafuge 40, SL40 and Eppendorf™ 5810/5804 Series, Sorvall Legend T+, Multifuge 3+ Series, Sorvall Legend 23R, MR23i, Thermo Scientific™ Nalgene™ bottles and tubes
Note:
†Performance may vary by centrifuge model.
Specifications
Fiberlite F21-48 x 2 Fixed Angle Rotor for Thermo Scientific Sorvall Legend X1, X3, Sorvall ST 40 | |
48 x 2 mL | |
15,200 rpm | |
455 | |
11 mm | |
45° | |
Yes |
Fixed Angle Rotor | |
25,055 x g | |
5.1 cm | |
50 mm | |
Thermo Scientific Sorvall Legend XT/XF, Sorvall Legend X1, Sorvall ST 40, Sorvall ST 16, Multifuge X3/X3F, Multifuge X1, Megafuge 40, Megafuge | |
Biocontainment lid | |
Energy efficient, less waste |