Learn More
Cytiva Ficoll-Paque™ PLUS Media
Recognized standard in laboratories worldwide for isolation of human lymphocytes from peripheral blood. Cytiva™ Ficoll-Paque™ PLUS Media maintains the viability and a representative distribution of B and T lymphocytes.
Brand: Cytiva 17-1440-03
518.70 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : A0
Additional Details : CAS Number : 156-57-0 Weight : 5.04000kg
Description
- Ready-to-use sterile medium
- Density media optimized for the isolation of lymphocytes from peripheral blood using simple centrifuge-based protocol
- High yield
- Maintains viability and representative distribution of B and T lymphocytes
- Low levels of endotoxin (<0.12 EU/mL)
Specifications
4–30°C | |
Cell Isolation | |
Colorless to Slightly Yellow | |
1.078 g/mL | |
Liquid |
6 x 500 mL | |
< 0.12 EU/mL | |
Centrifugation Media | |
Sterile |