missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Ficoll PM 70 Centrifugation Media
Behave as ideal neutral spheres. Cytiva™ Ficoll PM70 Centrifugation Media is considered the molecules of choice for studying pore size distribution and the permeability of membranes.
Brand: Cytiva 17-0310-10
This item is not returnable.
View return policy
Description
- High molecular weight (70,000) sucrose-polymer formed by copolymerization of sucrose with epichlorohydrin
- Highly branched structure and high content of hydroxyl groups mean good solubility in aqueous media with concentrations up to 50% w/v easily attained
- Displays minimal toxicity, plus favorable viscosity and osmotic properties
- Low endotoxin levels (<0.12 EU/mg)
For use in perfusion media for laboratory studies on dextran-sensitive animals

Specifications
100 g | |
Specifically intended for use in perfusion media for laboratory studies on dextran-sensitive animals. | |
0.12 EU/mg | |
Centrifugation Media |
4–30°C | |
≈10mL/g | |
White | |
Powder |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Ficoll PM 70 Centrifugation Media > 100g
Spot an opportunity for improvement?Share a Content Correction