missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Ficoll PM400
High molecular weight sucrose-polymers formed by copolymerization of sucrose with epichlorohydrin. Cytiva™ Ficoll PM400 is a synthetic neutral, highly-branched hydrophilic polymer of sucrose, used for producing density gradients for separation of cells and subcellular components by centrifugation or by sedimentation at unit gravity.
£165.36 - £4871.00
Specifications
Content And Storage | 4–30°C |
---|---|
For Use With (Application) | Cell Isolation |
Viscosity | 5 ηr at 20°C: 10% (w/v), 20 ηr at 20°C: 20% (w/v), 60 ηr at 20°C : 30% (w/v, 180 ηr at 20°C : 40% (w/v) , 600 ηr at 20°C : 50% (w/v) |
Endotoxin Level | 0.12 EU/mg |
Color | White |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11580724
|
Cytiva
17-0300-10 |
100 g |
£165.36
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11590724
|
Cytiva
17-0300-50 |
500 g |
£640.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11570724
|
Cytiva
17-0300-05 |
5 kg |
£4871.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For producing density gradients for separation of cells and subcellular components by centrifugation or by sedimentation at unit gravity
- Neutral, highly branched, hydrophilic polymer of sucrose which dissolves readily in aqueous solution
- Concentrations of up to 50% (w/v) covering densities of up to 1.2g/mL can be achieved
- Has better osmotic properties than sucrose
- Preserves functional and morphological integrity
- Useful for separating cells that are sensitive to centrifugation and for separating cells of similar density but different size
- Does not penetrate biological membranes
- Serves as the raw material for preparation of Ficoll-Paque gradients
- Used in other applications such as electrophoresis, hybridization, cryopreservation, and as a hapten carrier
- Supplied as dry powders
- Used in the formation of density gradients for the separation and isolation of eukaryotic cells, organelles, and bacterial cells, as a stabilizing agent and as a separation medium for the isolation of lymphocytes
- Used in defined culture media, nucleic acid hybridization, electrophoresis, and immunological studies
- Used for gradient centrifugation in all types of centrifuge rotors and for separation at unit gravity
Specifications
4–30°C | |
5 ηr at 20°C: 10% (w/v), 20 ηr at 20°C: 20% (w/v), 60 ηr at 20°C : 30% (w/v, 180 ηr at 20°C : 40% (w/v) , 600 ηr at 20°C : 50% (w/v) | |
White | |
1.2 g/mL |
Cell Isolation | |
0.12 EU/mg | |
Centrifugation Media | |
Powder |
Safety and Handling
missing translation for 'shelfLife' : 5 years
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva Ficoll PM400