missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Filter Holder

Polysulfone In-Line Filter Holder for 50mm Diameter Membrane

Brand:  Cytiva 10461300

Code : A0

Additional Details : Weight : 0.19000kg

 View more versions of this product

Product Code. 11359304

  • £80.70 / Each
Estimated Shipment: 19-07-2024
to see stock.
Add to basket

Description

Description

Polysulfone In-Line Filter Holder for 50mm Diameter Membrane

Specifications

Specifications

FP050/1 PSU
1
50 mm
Polysulfone
Filter Holder
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers