Learn More
Contec™ Filtered Alcohols: 70% IPA or Denatured Ethanol Blended with Purified Water
Ensures alcohol, container, packaging are cleanroom free from contamination and particulates.
Brand: Contec™ FBC570I
82.40 GBP valid until 2024-07-31
Use promo code "21674" to get your promotional price.
Additional Details : CAS Number : 67-63-0 Weight : 8.80000kg Transport : UN number : 1219 Chem class : 3 Pack group : II
Description
- GMP manufactured under Grade A air flow in a Grade C cleanroom
- Ensures the alcohol, container and packaging are free from contamination and particulates
- No denaturants other than IPA
- Completely residue free
- Trigger sprayer and bag in bottle protected system
- Prevents bottle contents from becoming contaminated during use
- No need to discard unused product during shelf life
- Bottles can be completely emptied eliminating wastage
- Trigger sprayer can be set to jet or spray
- Large droplet size reduces the risk of inhalation and provides good surface coverage
- Double bagged packed in linear tear packaging
- Each bag is easy to open even when wearing gloves
- Facilitates transfer disinfection into cleanroom
- 0.2 micron filtered, filled, bagged in Grade C (ISO Class7) cleanroom
- Endotoxin level: 0.25EU /mL
- No denaturants other than IPA
- Suitable for use in product contact areas
- Completely residue free
- Prevents bottle contents from becoming contaminated
- Facilitates transfer disinfection into high grade areas
- Double bagged in polyethylene linear tear bag
Specifications
Filtered Alcohol | |
Unscented | |
Colorless | |
Clear | |
70% Isopropanol in Purified Water | |
Limited-use | |
Yes | |
2 x 5 L |
Liquid | |
Sterile | |
Not Autoclavable | |
Isopropanol | |
Capped Bottle | |
Filtered | |
Sterilizes surfaces | |
5 L |
Safety and Handling
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use dry chemical, carbondioxide to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol