missing translation for 'onlineSavingsMsg'
Learn More

Ecolab Filtered Klercide 70/30 IPA Blended with Deionized Water

This Klercide 70/30 IPA blended with deionized water is cleanroom filtered for use in lower grade areas.

Brand:  Ecolab 6600949

Additional Details : CAS Number : 67-63-0 Weight : 5.26500kg Transport : UN number : 1219 Chem class : 3 Pack group : II


 View more versions of this product

Product Code. 17815497

  • £220.85 / 6 L
Estimated Shipment: 01-08-2024
to see stock.

Description

Description

  • BPR approved
  • Economical disinfection suitable for use in lower grade areas
  • 0.2 micron filtered, then filled and double bagged in a Grade C (ISO Class 7) cleanroom
  • Supplied in spray bottle which incorporates the patented SteriShield Delivery System and an adjustable trigger spray. The unique way in which the SteriShield Delivery System works, as a closed system, ensures that the integrity of the contents is preserved throughout use
  • Certificates of Analysis are available for every batch
  • Full supporting technical documentation is available
Specifications

Specifications

Disinfectant
Neutral
6 x 1 L
Liquid
Sterile
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Klercide 70/30 IPA

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

Supplemental information
  • MIXTURE LIST-Contains : Isopropanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers