missing translation for 'onlineSavingsMsg'
Learn More

Ecolab Filtered Klercide 70/30 IPA Blended with Deionized Water

This Klercide 70/30 IPA blended with deionized water is cleanroom filtered for use in lower grade areas.

£220.85 - £468.00

Specifications

Product Type Disinfectant
Form Liquid
pH Neutral
Sterility Sterile
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
17895487
View Documents Promotion Details
Ecolab
6600959
4 x 5 L
£468.00
20 L
View alternative products
Save Up to 
»
Estimated Shipment: 01-08-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
17815497
View Documents Promotion Details
Ecolab
6600949
6 x 1 L
£220.85
6 L
View alternative products
missing translation for 'toSaveUpto' 
»
Estimated Shipment: 01-08-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • BPR approved
  • Economical disinfection suitable for use in lower grade areas
  • 0.2 micron filtered, then filled and double bagged in a Grade C (ISO Class 7) cleanroom
  • Supplied in spray bottle which incorporates the patented SteriShield Delivery System and an adjustable trigger spray. The unique way in which the SteriShield Delivery System works, as a closed system, ensures that the integrity of the contents is preserved throughout use
  • Certificates of Analysis are available for every batch
  • Full supporting technical documentation is available
Specifications

Specifications

Disinfectant
Liquid
Neutral
Sterile
Videos
SDS
Documents

Documents

Certificates