Learn More
Ecolab Filtered Klercide 70/30 IPA Blended with Deionized Water
This Klercide 70/30 IPA blended with deionized water is cleanroom filtered for use in lower grade areas.
Brand: Ecolab 6600959
Additional Details : CAS Number : 67-63-0 Weight : 17.55000kg Transport : UN number : 1219 Chem class : 3 Pack group : II
Description
- BPR approved
- Economical disinfection suitable for use in lower grade areas
- 0.2 micron filtered, then filled and double bagged in a Grade C (ISO Class 7) cleanroom
- Supplied in spray bottle which incorporates the patented SteriShield Delivery System and an adjustable trigger spray. The unique way in which the SteriShield Delivery System works, as a closed system, ensures that the integrity of the contents is preserved throughout use
- Certificates of Analysis are available for every batch
- Full supporting technical documentation is available
Specifications
Disinfectant | |
Neutral | |
4 x 5 L |
Liquid | |
Sterile |