missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers

Multiskan™ FC microplate photometers can be equipped with wide range of additional filters to expand the instrument functionality and application flexibility.

Brand:  Thermo Scientific™ 1427505

Code : A0

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 11713869

  • £590.00 / Each
Estimated Shipment: 03-09-2024
to see stock.

Description

Description

Multiskan FC brings together over 40 years' experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility.

  • Multiskan FC is equipped with an eight-position filter wheel with three preinstalled standard filters, 405 nm, 450 nm and 620 m.
  • A comprehensive range of easy-to install additional filters is available for order
  • Superior usability and logical workflow with Thermo Scientific™ SkanIt™ Software
Specifications

Specifications

Optical Filter
Multiskan FC
Multiskan™
750 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers