Learn More
Thermo Scientific™ Filters and Accessories for Thermo Scientific Multiskan™ FC Microplate Readers
Multiskan™ FC microplate photometers can be equipped with wide range of additional filters to expand the instrument functionality and application flexibility.
£243.00 - £890.00
Product Code | Brand | Product Type | Wavelength | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Wavelength | Price | Quantity & Availability | ||||
11540705
|
Thermo Scientific™
1423755 |
Optical Filter | 375 nm |
£688.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11709458
|
Thermo Scientific™
1424055 |
Optical Filter | 405 nm |
£248.00
Each |
Estimated Shipment: 03-09-2024
Log in to see stock. |
||||
11719458
|
Thermo Scientific™
1424145 |
Optical Filter | 414 nm |
£248.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11729458
|
Thermo Scientific™
1424505 |
Optical Filter | 450 nm |
£249.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11799448
|
Thermo Scientific™
1424925 |
Optical Filter | 492 nm |
£244.00
Each |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12566528
|
Thermo Scientific™
1425205 |
Optical Filter | 520 nm |
£686.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11560705
|
Thermo Scientific™
1425305 |
Optical Filter | 530 nm |
£890.00
Each |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11755058
|
Thermo Scientific™
1425405 |
Optical Filter | 540 nm |
£244.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11709508
|
Thermo Scientific™
1425505 |
Optical Filter | 550 nm |
£244.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11761218
|
Thermo Scientific™
1425605 |
Optical Filter | 560 nm |
£712.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11794968
|
Thermo Scientific™
1425705 |
Optical Filter | 570 nm |
£244.00
Each |
Estimated Shipment: 17-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11829540
|
Thermo Scientific™
1425955 |
Optical Filter | 595 nm |
£244.00
Each |
Estimated Shipment: 27-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11500705
|
Thermo Scientific™
1426205 |
Optical Filter | 620 nm |
£249.00
Each |
Estimated Shipment: 29-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11530705
|
Thermo Scientific™
1426305 |
Optical Filter | 630 nm |
£480.00
Each |
Estimated Shipment: 11-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11765058
|
Thermo Scientific™
1426505 |
Optical Filter | 650 nm |
£244.00
Each |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11782278
|
Thermo Scientific™
1426905 |
Optical Filter | 690 nm |
£243.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11580705
|
Thermo Scientific™
1427405 |
Optical Filter | 740 nm |
£684.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11713869
|
Thermo Scientific™
1427505 |
Optical Filter | 750 nm |
£590.00
Each |
Estimated Shipment: 03-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11500715
|
Thermo Scientific™
1428205 |
Optical Filter | 820 nm |
£886.00
Each |
Estimated Shipment: 27-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Multiskan FC brings together over 40 years' experience of the well-known Multiskan product brand, with features for enhanced usability. The Multiskan FC has a 340–850 nm wavelength range, enabling a wide variety of applications from enzyme kinetic studies to Lowry assays. Extensive choice of filters for optimal wavelength selection for assays increases the application flexibility.
- Multiskan FC is equipped with an eight-position filter wheel with three preinstalled standard filters, 405 nm, 450 nm and 620 m.
- A comprehensive range of easy-to install additional filters is available for order
- Superior usability and logical workflow with Thermo Scientific™ SkanIt™ Software
Specifications
Optical Filter | |
Multiskan FC |