Learn More
Thermo Scientific™ Finnpipette™ Novus GLP Kits

Access everything needed for pipetting in these all-inclusive GLP kits featuring ergonomic and easy-to-use pipetters.
Brand: Thermo Scientific™ 4701120
1176.47 GBP valid until 2025-12-31
Use promo code "24294" to get your promotional price.
Description
Access everything needed for pipetting in these all-inclusive Thermo Scientific™ Finnpipette™ Novus GLP kits. The kits cover the volume range you need with four different options to choose from, and include Finnpipette™ Novus™ pipetters, appropriate stands and sample tips.
Novus GLP Kit 1 includes:
- Finnpipette Novus 10-100ÎĽL
- Finnpipette Novus 100-1000ÎĽL
- Two Finnpipette Novus stands
- One rack of Finntip Flex 200 and Flex 1000
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Pipetting Guide
- Finnpipette Novus brochure
Novus GLP Kit 2 includes:
- Finnpipette Novus 100-1000ÎĽL
- Finnpipette Novus 0.5-5mL
- Two Finnpipette Novus stands
- One rack of Finntip 5ml and Flex 1000
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Finnpipette Novus brochure
- Pipetting Guide
Novus GLP Kit 3 includes:
- Novus pipette, 1-10ÎĽL, micro
- Novus pipette, 10-100ÎĽL
- Novus pipette, 100-1000ÎĽL
- Three Finnpipette Novus pipette stands
- One rack of Flex 10, Flex 200 and Flex 1000
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Pipetting Guide
- Novus brochure
Novus GLP Kit 4 includes:
- Novus pipette, 10-100ÎĽL
- Novus pipette, 100-1000ÎĽL
- Novus pipette, 1-10mL
- Three Finnpipette Novus pipette stands
- One rack of Flex 200, Flex 1000 and Finntip 10ml
- Reagent Reservoirs demo pack
- Finntip Sample Pack
- Pipetting Guide
- Novus brochure

Specifications
Novus | |
Novus | |
1 to 1000 ÎĽL |
1 | |
GLP Kit | |
Novus 10micro, 100 and 1000 ÎĽL single channel pipettes, three Novus stands, three sample Finntip racks |
Product Suggestions
Customers who viewed this item also viewed.
Videos
-
Thermo Scientific Finnpipette Novus electronic pipettors
-
Thermo Scientific Finnpipette Novus electronic pipettors
Your input is important to us. Please complete this form to provide feedback related to the content on this product.