Learn More
Granulated Agar, Fisher BioReagents
Used as a solidifying agent to provide the solid medium to support the growth of microbes
Brand: Fisher Bioreagents BP9744-500
Code : NEW
Additional Details : Weight : 0.50000kg
Description
- Granulated agar media CAS:[9002-18-0]
- Store at room temperature
Specifications
Granulated Powder | |
Used as a solidifying agent to provide the solid medium to support the growth of microbes | |
500 g |
Agar | |
Poly Bottle |