missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granulated Agar, Fisher BioReagents
Used as a solidifying agent to provide the solid medium to support the growth of microbes
£96.29 - £728.00
Specifications
Form | Granulated Powder |
---|---|
Product Type | Agar |
For Use With (Application) | Used as a solidifying agent to provide the solid medium to support the growth of microbes |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
15480665
|
Fisher Bioreagents
BP9744-5 |
Poly Pail | 5 kg |
£728.00
5kg |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
15450665
|
Fisher Bioreagents
BP9744-500 |
Poly Bottle | 500 g |
£96.29
500g |
Estimated Shipment: 02-08-2024
Log in to see stock. |
||||
15470665
|
Fisher Bioreagents
BP9744-2 |
Poly Pail | 2 kg |
£328.18
2kg |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
Description
- Granulated agar media CAS:[9002-18-0]
- Store at room temperature
Specifications
Granulated Powder | |
Agar | |
Used as a solidifying agent to provide the solid medium to support the growth of microbes |