Learn More
Fisher BioReagents™ Fisher BioReagents™ Microbiology Media Additives: Acid Casein Peptone
Additive for preparing bacterial nutrient media
Brand:  Fisher BioReagents™ 12747109
 View more versions of this product
Product Code. 12747109
  • £88.60 / 500g
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
Acid Casein Peptone
Dehydrated.
Specifications
Specifications
Powder
Casamino Acid
Poly Bottle
Molecular genetics
Acid hydrolyzed casein's low concentrations of sodium chloride and iron permit toxin production
500 g