missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Microbiology Media Additives: Casamino Acids
Additive for preparing bacterial nutrient media
£81.33 - £207.76
Specifications
Form | Powder |
---|---|
Grade | Molecular genetics |
Product Type | Casamino Acid |
For Use With (Application) | Acid hydrolyzed casein's low concentrations of sodium chloride and iron permit toxin production |
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging Type | Quantity | Price | Quantity & Availability | ||||
11493853
|
Fisher BioReagents™
11493853 |
Poly Pail | 2000 g |
£207.76
2kg |
Estimated Shipment: 30-08-2024
Log in to see stock. |
||||
12747109
|
Fisher BioReagents™
12747109 |
Poly Bottle | 500 g |
£81.33
500g |
In Stock
Log in to see stock. |
||||
Description
Dehydrated.
Specifications
Powder | |
Molecular genetics | |
Casamino Acid | |
Acid hydrolyzed casein's low concentrations of sodium chloride and iron permit toxin production |