missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Microbiology Media: LB Broth (Powder) - Lennox
Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.
Brand: Fisher BioReagents™ BP1427-500
This item is not returnable.
View return policy
Description
- Directions for Media Preparation: Suspend 20g in 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve. Before use, sterilize by autoclaving for 15 minutes
- Provides a nutrient-rich environment to enable faster growth of E. coli
- Format: Dehydrated powder
Specifications
Bacterial Culture | |
Molecular genetics | |
Microbiology Media | |
Preparing bacterial nutrient media | |
500 g |
Powder | |
LB Broth - Lennox | |
Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 5g/L | |
Poly Bottle |
Safety and Handling
Product Identifier
- Broth Miller
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisher BioReagents™ Microbiology Media: LB Broth (Powder) - Lennox > LB Broth; 500g
Spot an opportunity for improvement?Share a Content Correction