missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ Microbiology Media: LB Broth (Powder) - Lennox

Used as a microbial growth medium for the cultivation of E. coli strains for the preparation of plasmid DNA and recombinant proteins.

£57.67 - £167.96

Specifications

Description Bacterial Culture
Form Powder
Grade Molecular genetics
Media Type LB Broth - Lennox
Product Type Microbiology Media
View More Specs

Products 2
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
Product Code Brand Quantity Packaging Type Price Quantity & Availability  
10628973
View Documents
Fisher BioReagents™
BP1427-500
500 g Poly Bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10285103
View Documents
Fisher BioReagents™
BP1427-2
2000 g Poly Pail This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Directions for Media Preparation: Suspend 20g in 1L of purified water, such as BP2820 (Fisher BioReagents Microbial Cell Culture Grade Water). Heat to completely dissolve. Before use, sterilize by autoclaving for 15 minutes
  • Provides a nutrient-rich environment to enable faster growth of E. coli
  • Format: Dehydrated powder
Specifications

Specifications

Bacterial Culture
Molecular genetics
Microbiology Media
Preparing bacterial nutrient media
Powder
LB Broth - Lennox
Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 5g/L
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Broth Miller

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisher BioReagents™ Microbiology Media: LB Broth (Powder) - Lennox

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.