missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 150 and 850 Hand Held Homogenizer Accessory, Stainless Steel Probes
442.75 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Alert:
Warning: Stainles Steel bearings cannot be autoclaved.
Description
- Provides end user flexibility to process samples
- Designed for long life and are easy to disassemble for cleaning
- Stator design allows fine to medium samples to pass through the processing windows
- Comes with PTFE upper and lower bearings
- Autoclavable
Specifications
Probe | |
0.25 to 30 mL | |
4.5 in. | |
Stainless Steel | |
150 Hand Held Homogenizer and 850 Homogenizers |
Course | |
7 mm | |
115 mm | |
Sawtooth, wide window |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction