missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 850 Homogenizer Accessory, Generator Probe
Stainless-steel generator probe; Fisher Scientific™ 850 Homogenizer Accessory, Generator Probe is compatible with Fisher Scientific 850 Homogenizers.
Brand: Fisherbrand™ 15515829
Alert:
Warning: Stainles Steel bearings cannot be autoclaved.
Includes: Generator probe, instruction manual, cleaning brush and spare bearing
Description
- Comes with PTFE upper and lower bearings
- Entire probe can be autoclaved
- Easily disassembled for cleaning
Saw Tooth Generator Probe:
- Add extra cutting action and stator design allows fine to medium samples to pass through processing windows
Specifications
Probe | |
50 mL to 2 L | |
195 mm | |
Sawtooth |
Fine | |
20 mm | |
316 Stainless Steel | |
Fisher Scientific 850 Homogenizer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction