missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ accumet™ Liquid-Filled Mercury-Free pH/ATC Epoxy Body Combination Electrodes, w/BNC and Mini-phone ATC connectors
TRIS compatible, rugged, and convenient
Brand: Fisherbrand™ 01X417061
136.33 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Includes: Electrode, 60mL refill solution, electrode storage bottle
Requires : Requires filling solution
Description
TRIS compatible, rugged, and convenient
- Rugged standard-size epoxy body
- Double-junction design prevents reference contamination; compatible with TRIS applications; mercury free
- Liquid filled design provides quick response
- Porous ceramic junction, Ag/AgCl reference
- Built-in ATC probe
- Rugged epoxy body for use in field or plant
- Ideal for handheld use with aqueous samples
- Double-junction design eliminates measurement problems and clogging in research with TRIS buffers
Order Info
Endcap seal; keeps membrane moist and ready
Specifications
Purple | |
Porous Ceramic Double | |
0 to 14 | |
pH/ATC Combination Titration Electrode | |
General laboratory and life science applications | |
Electrode, 60mL refill solution, electrode storage bottle | |
ATC | |
0°C to 80°C | |
106 mm | |
12 mm |
BNC/Mini-Phone ATC | |
Epoxy | |
0°C to 80°C | |
Saturated KCl Electrolyte (SP138-500) | |
Accumet Excel (XL) series, Research (AR) and Basic (AB15 and AB15 Plus) benchtop meter | |
Ag/AgCl | |
32°F to 212°F | |
4.1 in. | |
0.47 in. |