Learn More
Fisherbrand™ Maxima C Plus Centering Rings
Centering Ring for use with Maxima C vacuum pumps to join two flanges
£10.25 - £25.98
Product Code | Brand | For Use With | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | ||||||
10243801
|
Fisherbrand™
FB70127 |
Model M2C and M4C vacuum pump | N/A | |||||||
10534002
|
Fisherbrand™
701073 |
Model M8C, M12C and M16C vacuum pump |
£10.25
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
Description
For use with Model M2C and M4C vacuum pumpSpecifications
Stainless Steel | |
Centering Ring |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Maxima C Plus Centering Rings